Learn More
Microsolv Technology Corporation Tubing Holder, Knee, 90 Degree Turn. Blue. 10/PK.
Supplier:  Microsolv Technology Corporation 4655290
Encompass_Preferred
This Tubing Holder, will keep your tubing "in place" when making a 90⁰ turn, which is not easy. Avoid clutter and keep your system optimized. These tubing knees will give you the right bend of your PEEK tubing, preventing close diameters, which do have a negative effect on the mechanical stability of your PEEK tubing and can cause poor internal dimensions.Best performance when used with 1/16th" PEEK Tubing but other tubing can be used as well.
Catalog No. 50-227-1445