Learn More
Microsolv Technology Corporation Tubing Holder, Knee, 180 Degree Turn. Blue. 10/PK.
Catalog No. 502271444
Encompass_Preferred
This Tubing Holder, will keep your tubing "in place" when making a 180⁰ turn, which is not always easy. Avoid clutter and keep your system optimized. These tubing knees will give you the right bend of your PEEK tubing, preventing close diameters, which do have a negative effect on the mechanical stability of your PEEK tubing and can cause poor internal dimensions.Best performance when used with 1/16th" PEEK Tubing but other tubing can be used as well.
This item is not returnable. View return policy
Catalog No. 50-227-1444 Supplier Microsolv Technology Corporation Supplier No. 46552180
This Tubing Holder, will keep your tubing "in place" when making a 180⁰ turn, which is not always easy. Avoid clutter and keep your system optimized. These tubing knees will give you the right bend of your PEEK tubing, preventing close diameters, which do have a negative effect on the mechanical stability of your PEEK tubing and can cause poor internal dimensions.Best performance when used with 1/16th" PEEK Tubing but other tubing can be used as well.