Learn More
Waters Corp Waters HPLC Tubing, Stainless Steel, .005 in ID, 24 in Long
Catalog No. 50791942 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC inlet tubing is a stainless steel assembly specifically designed for use with the ACQUITY 30cm Column Heater/Cooler. With an internal diameter of .005 in and a length of 24 in, this tubing connects the column to inject valve port 6 and is essential for performing AQT testing.
  • Made from stainless steel for durability and chemical resistance.
  • Specifically designed as an inlet tube for ACQUITY 30cm Column Heater/Cooler.
  • Essential for performing AQT testing.
  • Connects the column to the inject valve port 6.
  • Ideal for high-performance liquid chromatography applications.
Catalog No. 50-791-942 Supplier Waters Corp Supplier No. 430001923
This HPLC inlet tubing is a stainless steel assembly specifically designed for use with the ACQUITY 30cm Column Heater/Cooler. With an internal diameter of .005 in and a length of 24 in, this tubing connects the column to inject valve port 6 and is essential for performing AQT testing.
  • Made from stainless steel for durability and chemical resistance.
  • Specifically designed as an inlet tube for ACQUITY 30cm Column Heater/Cooler.
  • Essential for performing AQT testing.
  • Connects the column to the inject valve port 6.
  • Ideal for high-performance liquid chromatography applications.