Learn More
Accuflow Systems Saint-Gobain Peristaltic Tubing, Platinum-Cured Silicone, 0.250/0.500 in (ID/OD)
Catalog No. NC3255262
Encompass_Preferred
Sani-Tech SPT-60L is a pharmaceutical-grade, platinum-cured silicone tubing engineered for high-precision fluid transfer in peristaltic pumps and final-fill operations. Controlled dimensions and low spallation deliver accurate dosing and extended pump life for biopharmaceutical manufacturing and single-use assemblies.
  • Platinum-cured silicone for low extractables and bioprocess compatibility.
  • 0.250 in inner diameter, 0.500 in outer diameter, 0.125 in wall thickness.
  • Nominal 60 Shore A durometer for consistent pump performance.
  • Engineered for low spallation and extended peristaltic pump life.
  • Suitable for final-fill and peristaltic dosing applications.
  • Compliance: USP <88> Class VI, USP <87>, ISO 10993-5; see datasheet for full validation details.
Catalog No. NC3255262 Supplier Accuflow Systems Supplier No. SPT60L2504
Sani-Tech SPT-60L is a pharmaceutical-grade, platinum-cured silicone tubing engineered for high-precision fluid transfer in peristaltic pumps and final-fill operations. Controlled dimensions and low spallation deliver accurate dosing and extended pump life for biopharmaceutical manufacturing and single-use assemblies.
  • Platinum-cured silicone for low extractables and bioprocess compatibility.
  • 0.250 in inner diameter, 0.500 in outer diameter, 0.125 in wall thickness.
  • Nominal 60 Shore A durometer for consistent pump performance.
  • Engineered for low spallation and extended peristaltic pump life.
  • Suitable for final-fill and peristaltic dosing applications.
  • Compliance: USP <88> Class VI, USP <87>, ISO 10993-5; see datasheet for full validation details.