Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Tubing, Inlet to Diverter, 0.040 in.

Catalog No. 502390786 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This tubing is specifically designed to function as an inlet to a diverter, ensuring precise flow control in various applications. It features a specific internal diameter to facilitate optimal performance in compatible systems.

  • Functions as an inlet to a diverter.
  • Features a 0.040 in. internal diameter.
  • Weighs approximately 50 g.
  • Manufactured in the U.S.A.

Catalog No. 50-239-0786 Supplier Waters Corp Supplier No. 700001657
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This tubing is specifically designed to function as an inlet to a diverter, ensuring precise flow control in various applications. It features a specific internal diameter to facilitate optimal performance in compatible systems.

  • Functions as an inlet to a diverter.
  • Features a 0.040 in. internal diameter.
  • Weighs approximately 50 g.
  • Manufactured in the U.S.A.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.