Learn More
Julabo USA Inc 1m tubing insulation for 8mm and 10mm ID tubing
SDP
Catalog No. 507534148
Encompass_Preferred
Bath Accessories, Julabo, 1 m Insulation with 14 mm inner diameter, for rubber tubing 8 to 10 mm ID. For use with 8930008, 8930010, 8930108, 8930110. Priced per meter.
Catalog No. 50-753-4148 Supplier Julabo USA Inc Supplier No. 8930410
Bath Accessories, Julabo, 1 m Insulation with 14 mm inner diameter, for rubber tubing 8 to 10 mm ID. For use with 8930008, 8930010, 8930108, 8930110. Priced per meter.