Learn More
Waters Corp Tubing Kit, 4-Column Bank Connection, CM-S
Catalog No. 502390680 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit is designed for a 4-column bank connection.
  • Designed for 4-column bank connections.
  • Includes 2 units of tube assembly, 0.005 in. ID, for inline column to column connections.
  • Includes 2 units of tube assembly, 0.004 in. ID, for aligned, vertical, column to column connections.
  • Includes 1 unit of tube assembly, 0.004 in. ID, for offset, vertical, column to column connections.
Catalog No. 50-239-0680 Supplier Waters Corp Supplier No. 205001172
This tubing kit is designed for a 4-column bank connection.
  • Designed for 4-column bank connections.
  • Includes 2 units of tube assembly, 0.005 in. ID, for inline column to column connections.
  • Includes 2 units of tube assembly, 0.004 in. ID, for aligned, vertical, column to column connections.
  • Includes 1 unit of tube assembly, 0.004 in. ID, for offset, vertical, column to column connections.