Learn More
Waters Corp Tubing Kit, CH-30A, MP35N
Supplier:  Waters Corp 205000792
Encompass_Preferred
This tubing kit is designed for use with the ACQUITY UPLC Column Heater-30A within the ACQUITY UPLC H-Class Bio and ACQUITY UPLC H-Class PLUS Bio Systems. It contains various tubes that connect different components within the UPLC system, facilitating the flow path for the mobile phase.
  • Contains tubes for connecting pre-heaters to columns.
  • Includes tubes for connecting columns to detectors.
  • Utilizes MP35N material for key components.
  • Features Pharmed tubing for improved insulation against environmental temperature changes.
  • Specifically designed for the 30-cm Column Heater-Active.
Catalog No. 50-239-2326