Learn More
Waters Corp Tubing Configuration Kit, ACQUITY Advanced Polymer Chromatography, No Valves, CM-S
Catalog No. 502392088 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides tubing and accessories for configuring an ACQUITY Advanced Polymer Chromatography (APC) system. It is designed for use with the ACQUITY APC Single-Zone Column Manager (CM-S) in configurations without valves. The kit includes various tubing assemblies for column connections and detector interfaces, along with components for column retention and fluidic connection identification.
  • Designed for advanced polymer chromatography systems.
  • Configured for single-zone column managers without valves.
  • Includes various tubing assemblies for column connections and detector interfaces.
  • Contains components for column retention.
  • Features tube markers for identifying fluidic connections.
  • Includes a magnetic label for ease of plumbing.
  • Supplied with spare ferrules for reusable fittings.
Catalog No. 50-239-2088 Supplier Waters Corp Supplier No. 205001166
This kit provides tubing and accessories for configuring an ACQUITY Advanced Polymer Chromatography (APC) system. It is designed for use with the ACQUITY APC Single-Zone Column Manager (CM-S) in configurations without valves. The kit includes various tubing assemblies for column connections and detector interfaces, along with components for column retention and fluidic connection identification.
  • Designed for advanced polymer chromatography systems.
  • Configured for single-zone column managers without valves.
  • Includes various tubing assemblies for column connections and detector interfaces.
  • Contains components for column retention.
  • Features tube markers for identifying fluidic connections.
  • Includes a magnetic label for ease of plumbing.
  • Supplied with spare ferrules for reusable fittings.