Learn More
Perkin Elmer US LLC Peristaltic PESI Pump Tubing Kit - Qty.12
Catalog No. 509042309
Encompass_Preferred
The Peristaltic PESI Pump Tubing Kit includes two-stop tygon tubing with an inner diameter of 2.06 mm and a wall thickness of 0.80 mm. This kit ships with the Peristaltic Pump accessory. With this part number, this kit can be ordered as a replacement or spare. Qty. 12 tubes
Catalog No. 50-904-2309 Supplier Perkin Elmer US LLC Supplier No. B0190034
The Peristaltic PESI Pump Tubing Kit includes two-stop tygon tubing with an inner diameter of 2.06 mm and a wall thickness of 0.80 mm. This kit ships with the Peristaltic Pump accessory. With this part number, this kit can be ordered as a replacement or spare. Qty. 12 tubes