Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Precision Pump Tubing, Norprene Food, I/P 88; 25 ft
Catalog No. NC1820188
Encompass
Masterflex Norprene Food High-Performance Precision pump tubing is formulated to withstand extreme temperatures and will not deteriorate in demanding food and beverage applications. This tubing is compatible with virtually all commercial clears and sanitizers and can be sterilized via autoclave without impacted overall service life. Ideal for food and beverage applications, this tubing complies with FDA, NSF, and 3-A standards.
This item is not returnable. View return policy
Catalog No. NC1820188 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640288
Masterflex Norprene Food High-Performance Precision pump tubing is formulated to withstand extreme temperatures and will not deteriorate in demanding food and beverage applications. This tubing is compatible with virtually all commercial clears and sanitizers and can be sterilized via autoclave without impacted overall service life. Ideal for food and beverage applications, this tubing complies with FDA, NSF, and 3-A standards.