Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Sanitary Fitting High-Performance Precision Pump Tubing Assembly, PharMed BPT, L/S 24; 10 ft
Catalog No. NC9465222
Encompass
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Requires sanitary gaskets for both connections to ensure quality seal
This item is not returnable. View return policy
Catalog No. NC9465222 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9610324
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Requires sanitary gaskets for both connections to ensure quality seal