Learn More
Accuflow Systems TUBING MFLEX C-FLEX #16 25FT
Catalog No. 501108823
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex, C-Flex (50 A), L/S 16, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable, weldable Economical tubing Applica
Catalog No. 50-110-8823 Supplier Accuflow Systems Supplier No. 0642416
Manufactured by Cole Parmer and sold by Accuflow. Masterflex, C-Flex (50 A), L/S 16, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable, weldable Economical tubing Applica