Learn More
Accuflow Systems TUBING MFLEX C-FLEX #24 25FT
Catalog No. 501108857
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing (50 A), L/S 24, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable and weldable Economical tubing A
Catalog No. 50-110-8857 Supplier Accuflow Systems Supplier No. 0642424
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing (50 A), L/S 24, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable and weldable Economical tubing A