Learn More
Accuflow Systems TUBING MFLEX C-FLEX #24 400FT
Supplier:  Accuflow Systems 0642724
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex,C-Flex spooled, L/S 24, 400 ft. FDA compliant Optically inspected and factory tested. Ensures that your tubing is clean, free from defects, and performs to exact specifications. Thirteen tubing
Catalog No. 50-110-8973