Learn More
Accuflow Systems TUBING MFLEX C-FLEX #25 25FT
Catalog No. 501108837
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex, C-Flex (50 A), L/S 25, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable, weldable Economical tubing Applicatio
Catalog No. 50-110-8837 Supplier Accuflow Systems Supplier No. 0642425
Manufactured by Cole Parmer and sold by Accuflow. Masterflex, C-Flex (50 A), L/S 25, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable, weldable Economical tubing Applicatio