Learn More
Accuflow Systems TUBING MFLEX C-FLEX #36 25FT
Catalog No. 501108830
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing (50 A), L/S 36, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable and weldable Economical tubin
Catalog No. 50-110-8830 Supplier Accuflow Systems Supplier No. 0642436
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing (50 A), L/S 36, 25 ft. Combines biocompatibility of silicone with chemical resistance similar to Tygon Very low protein binding Heat sealable and weldable Economical tubin