Learn More
Accuflow Systems TUBING MFLEX C-FLEX #73 100FT
Supplier:  Accuflow Systems 0642773
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. I/P C-Flex spooled tubing, I/P 73, 100 ft per spool. Long tubing life Ultra smooth inner surface Fewer extractables Similar to silicone with chemical compatibility of Tygon Nontoxic; no leachable plas
Catalog No. 50-110-9025