Learn More
Accuflow Systems TUBING MFLEX C-FLEX #73 25FT
Catalog No. 501108850
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing (50 A), I/P 73, 25 ft. Applications: General-purpose lab uses requiring high performance and long life. Characteristics: Exceptional tensile and tear strength; good biocompatibili
Catalog No. 50-110-8850 Supplier Accuflow Systems Supplier No. 0642473
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing (50 A), I/P 73, 25 ft. Applications: General-purpose lab uses requiring high performance and long life. Characteristics: Exceptional tensile and tear strength; good biocompatibili