Learn More
Accuflow Systems TUBING MFLEX C-FLEX #82 100FT
Catalog No. 501108936
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. I/P C-Flex spooled tubing, I/P 82, 100 ft per spool. Long tubing life Ultra smooth inner surface Fewer extractables Similar to silicone with chemical compatibility of Tygon Nontoxic; no leachable plas
Catalog No. 50-110-8936 Supplier Accuflow Systems Supplier No. 0642782
Manufactured by Cole Parmer and sold by Accuflow. I/P C-Flex spooled tubing, I/P 82, 100 ft per spool. Long tubing life Ultra smooth inner surface Fewer extractables Similar to silicone with chemical compatibility of Tygon Nontoxic; no leachable plas