Learn More
Accuflow Systems TUBING MFLEX E-FOOD #17.50FT
Catalog No. 501108868
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tygon E-Food (B-44-4X) tubing, L/S 17, 50 ft. Applications: Food and beverage processing and dispensing. Characteristics: Nonwetting, odorless, tasteless, and nontoxic. Transparent.
Catalog No. 50-110-8868 Supplier Accuflow Systems Supplier No. 0641817
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tygon E-Food (B-44-4X) tubing, L/S 17, 50 ft. Applications: Food and beverage processing and dispensing. Characteristics: Nonwetting, odorless, tasteless, and nontoxic. Transparent.