Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBING MFLEX FDA VITON 24 25FT
Catalog No. 502741994
Encompass
Masterflex L/S High-Performance Precision Pump Tubing, FDA-Compliant Viton, L/S 24; 25 ft, Avantor
Catalog No. 50-274-1994 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9641224
Masterflex L/S High-Performance Precision Pump Tubing, FDA-Compliant Viton, L/S 24; 25 ft, Avantor