Learn More
Accuflow Systems TUBING MFLEX NORPRENE #15 50FT
Catalog No. 501108874
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Norprene tubing (A 60 G), L/S 15, 50 ft. Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance. Characteristics: Nonconductive, heat-sea
Catalog No. 50-110-8874 Supplier Accuflow Systems Supplier No. 0640415
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Norprene tubing (A 60 G), L/S 15, 50 ft. Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance. Characteristics: Nonconductive, heat-sea