Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBING MFLEX NORPRENE 36 50FT
Catalog No. 502742965
Encompass
Masterflex L/S High-Performance Precision Pump Tubing, Versilon™ A-60-N, L/S 36; 50 ft, Avantor
Catalog No. 50-274-2965 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640436
Masterflex L/S High-Performance Precision Pump Tubing, Versilon™ A-60-N, L/S 36; 50 ft, Avantor