Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled High-Performance Precision Pump Tubing, PharMed BPT, L/S 36; 100 ft
Catalog No. NC0091968
Encompass
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC0091968 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9568736/DEL
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.