Learn More
Accuflow Systems TUBING MFLEX PHARMED #14 25FT
Catalog No. 501108890
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Tubing, L/S #14, 25' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extremely
Catalog No. 50-110-8890 Supplier Accuflow Systems Supplier No. 0650814
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Tubing, L/S #14, 25' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extremely