Learn More
Accuflow Systems TUBING MFLEX PHARMED #15 400FT
Catalog No. 501108992
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Spooled Tubing, L/S #15, 400' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extr
This item is not returnable. View return policy
Catalog No. 50-110-8992 Supplier Accuflow Systems Supplier No. 9568715
Add to Cart
Add to Cart
Request a Quote
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Spooled Tubing, L/S #15, 400' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extr