Learn More
Accuflow Systems TUBING MFLEX PHARMED #16 25FT
Catalog No. 501108899
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Tubing, L/S #16, 25' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extremely low
Catalog No. 50-110-8899 Supplier Accuflow Systems Supplier No. 0650816
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Tubing, L/S #16, 25' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extremely low