Learn More
Accuflow Systems TUBING MFLEX PHARMED #24 300FT
Supplier:  Accuflow Systems 9568724
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Spooled Tubing, L/S #24, 300' Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extr
Catalog No. 50-110-8993
Add to Cart
Request a Quote