Learn More
Accuflow Systems TUBING MFLEX PHARMED #73 25FT
Catalog No. 501108943
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Tubing, I/P #73, 25 ft. Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extremely
Catalog No. 50-110-8943 Supplier Accuflow Systems Supplier No. 0650873
Manufactured by Cole Parmer and sold by Accuflow. Masterflex PharMed BPT Tubing, I/P #73, 25 ft. Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Characteristics: Nontoxic; nonhemolytic; extremely