Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex B/T PerfectPosition™ Pump Tubing, PharMed BPT, B/T 91; 25 ft
Catalog No. NC0632863
Encompass
Application suitability: Suitable for use with Acids, Alkalis, Pressure, Vacuum, Viscous fluid, and Sterile Fluids. Not Recommended for use with organic solvents. Physical characteristics : Thermoplastic elastomer, polypropylene-based material with USP mineral oil. Excellent tensile strength, firm (stiff) material. Opaque, beige. Temperature range: (static): -51 to 132C (-60 to 270F) (dynamic): -20 to 100C (-4 to 212F) Meets classifications: USP Class VI European Pharmacopoeia (EP 3.2.9) Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Sterilize: EtO, Autoclave, Gamma Radiation
Catalog No. NC0632863 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0650891/DEL
Application suitability: Suitable for use with Acids, Alkalis, Pressure, Vacuum, Viscous fluid, and Sterile Fluids. Not Recommended for use with organic solvents. Physical characteristics : Thermoplastic elastomer, polypropylene-based material with USP mineral oil. Excellent tensile strength, firm (stiff) material. Opaque, beige. Temperature range: (static): -51 to 132C (-60 to 270F) (dynamic): -20 to 100C (-4 to 212F) Meets classifications: USP Class VI European Pharmacopoeia (EP 3.2.9) Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. Sterilize: EtO, Autoclave, Gamma Radiation