Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBING MFLEX PHARMED NO.90 25F
Catalog No. 502742808
Encompass
Masterflex B/T PerfectPosition Pump Tubing, PharMed BPT, B/T 90; 25 ft, Avantor
Catalog No. 50-274-2808 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0650890
Masterflex B/T PerfectPosition Pump Tubing, PharMed BPT, B/T 90; 25 ft, Avantor