Learn More
Accuflow Systems TUBING MFLEX SIL. #17 25FT
Supplier:  Accuflow Systems 9644017
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex BioPharm Plus platinum-cured silicone pump tubing, L/S 17, 25 ft. Provides longer life than other platinum & peroxide silicone tubing formulations Exceeds USP Class VI & 3A sanitary standards Idea
Catalog No. 50-110-8914