Learn More
Accuflow Systems TUBING MFLEX SIL. #18 400FT
Catalog No. 501109147
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex BioPharm silicone tubing, L/S 18, 400 ft. Platinum-Cured silicone Ultra-smooth inner surface Fewer extractables Minimizes particle entrapment Perfect for lab, biotech
This item is not returnable. View return policy
Catalog No. 50-110-9147 Supplier Accuflow Systems Supplier No. 9642318
Add to Cart
Add to Cart
Request a Quote
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex BioPharm silicone tubing, L/S 18, 400 ft. Platinum-Cured silicone Ultra-smooth inner surface Fewer extractables Minimizes particle entrapment Perfect for lab, biotech