Learn More
Accuflow Systems TUBING MFLEX SIL. #24 25FT
Catalog No. 501108932
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex BioPharm platinum-cured silicone pump tubing, L/S 24, 25 ft. Formulation: Tygon 3350 silicone, platinum cured. Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.
Catalog No. 50-110-8932 Supplier Accuflow Systems Supplier No. 9642024
Manufactured by Cole Parmer and sold by Accuflow. Masterflex BioPharm platinum-cured silicone pump tubing, L/S 24, 25 ft. Formulation: Tygon 3350 silicone, platinum cured. Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.