Learn More
Accuflow Systems TUBING MFLEX SIL. #26 25FT
Catalog No. 501108889
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 26, 25 ft. Ensure optimal performance from your Masterflex pump Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower protein bindin
Catalog No. 50-110-8889 Supplier Accuflow Systems Supplier No. 9641026
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 26, 25 ft. Ensure optimal performance from your Masterflex pump Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower protein bindin