Learn More
Accuflow Systems TUBING MFLEX SIL. #89 10FT
Catalog No. 501108818
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 89, 10 ft.; Nontoxic and nonhemolytic with excellent biocompatibility. Long pumping life; pump at continuous pressures up to 40 psi; low extractables and
Catalog No. 50-110-8818 Supplier Accuflow Systems Supplier No. 9641089
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 89, 10 ft.; Nontoxic and nonhemolytic with excellent biocompatibility. Long pumping life; pump at continuous pressures up to 40 psi; low extractables and