Learn More
Accuflow Systems TUBING MFLEX SIL. #91 10FT
Catalog No. 501109251
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Peroxide-Cured Tubing, B/T 91, 10' No additives, plasticizers or DOP; odorless & nontoxic, fungus-resistant Extremely good at low temperatures Weather, ozone, corona, radiation resistant Minimal t
Catalog No. 50-110-9251 Supplier Accuflow Systems Supplier No. 9640091
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Peroxide-Cured Tubing, B/T 91, 10' No additives, plasticizers or DOP; odorless & nontoxic, fungus-resistant Extremely good at low temperatures Weather, ozone, corona, radiation resistant Minimal t