Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBING MFLEX SILICONE 15 20/CS
Catalog No. 502742011
Encompass
Masterflex L/S Bulk-Packed High-Performance Precision Pump Tubing, Platinum-Cured Silicone, L/S 15; 20 Bags of 25 ft/Bg, Avantor
Catalog No. 50-274-2011 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640415
Masterflex L/S Bulk-Packed High-Performance Precision Pump Tubing, Platinum-Cured Silicone, L/S 15; 20 Bags of 25 ft/Bg, Avantor