Learn More
Accuflow Systems TUBING MFLEX SILICONE #15 25FT
Catalog No. 501108923
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex BioPharm platinum-cured silicone pump tubing, L/S 15, 25 ft. Formulation: Tygon 3350 silicone, platinum cured. Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.
Catalog No. 50-110-8923 Supplier Accuflow Systems Supplier No. 9642015
Manufactured by Cole Parmer and sold by Accuflow. Masterflex BioPharm platinum-cured silicone pump tubing, L/S 15, 25 ft. Formulation: Tygon 3350 silicone, platinum cured. Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.