Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, L/S 15; 400 ft
Catalog No. NC0151201
Encompass
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC0151201 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9642315
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.