Learn More
Accuflow Systems TUBING MFLEX SILICONE #16 25FT
Catalog No. 501108854
Encompass_Preferred
Masterflex peroxide-cured silicone tubing, L/S 16, 25 ft. Ensure optimal performance from your Masterflex® pump Greater physical compression capability Economical, longer tubing life Masterflex® L/S® pump tubing is manufactured to extremely close tolerances that match our L/S® pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.
Catalog No. 50-110-8854 Supplier Accuflow Systems Supplier No. 9640016
Masterflex peroxide-cured silicone tubing, L/S 16, 25 ft. Ensure optimal performance from your Masterflex® pump Greater physical compression capability Economical, longer tubing life Masterflex® L/S® pump tubing is manufactured to extremely close tolerances that match our L/S® pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.