Learn More
Accuflow Systems TUBING MFLEX SILICONE #36 25FT
Catalog No. 501108921
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex peroxide-cured silicone tubing, L/S 36, 25 ft. Ensure optimal performance from your Masterflex pump Greater physical compression capability Economical, longer tubing life Applications: Pharm
Catalog No. 50-110-8921 Supplier Accuflow Systems Supplier No. 9640036
Manufactured by Cole Parmer & sold by Accuflow. Masterflex peroxide-cured silicone tubing, L/S 36, 25 ft. Ensure optimal performance from your Masterflex pump Greater physical compression capability Economical, longer tubing life Applications: Pharm