Learn More
Accuflow Systems TUBING MFLEX SILICONE #70 10FT
Catalog No. 501109234
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 70, 10 ft. 96510-70. Nontoxic and nonhemolytic with excellent biocompatibility Long pumping life; pump at continuous pressures up to 40 psi Low extractables a
Catalog No. 50-110-9234 Supplier Accuflow Systems Supplier No. 9651070
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 70, 10 ft. 96510-70. Nontoxic and nonhemolytic with excellent biocompatibility Long pumping life; pump at continuous pressures up to 40 psi Low extractables a