Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBING MFLEX SILICONE 70 10FT
Catalog No. 502741883
Encompass
Masterflex I/P High-Performance Precision Pump Tubing, Platinum-Cured Silicone, I/P 70; 10 ft, Avantor
Catalog No. 50-274-1883 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9651070
Masterflex I/P High-Performance Precision Pump Tubing, Platinum-Cured Silicone, I/P 70; 10 ft, Avantor