Learn More
Accuflow Systems TUBING MFLEX SILICONE #73 25FT
Catalog No. 501108952
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex BioPharm Plus platinum-cured silicone pump tubing, I/P 73, 25 ft. Specifications: Pump Series - I/P Masterflex tubing size (I/P) - 73 Tubing ID (mm) - 9.5 Hose barb size (in.) - 3/8 Flow range wit
Catalog No. 50-110-8952 Supplier Accuflow Systems Supplier No. 9644073
Manufactured by Cole Parmer & sold by Accuflow. Masterflex BioPharm Plus platinum-cured silicone pump tubing, I/P 73, 25 ft. Specifications: Pump Series - I/P Masterflex tubing size (I/P) - 73 Tubing ID (mm) - 9.5 Hose barb size (in.) - 3/8 Flow range wit