Learn More
Accuflow Systems TUBING MFLEX SILICONE #87 10FT
Catalog No. 501109263
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Platinum-Cured Silicone Tubing, B/T 87, 10' No leachable additives, DOP, or plasticizers; phthalate & latex-free; odorless & nontoxic, fungus, weather, ozone, corona, & radiation-resistant Extreme
Catalog No. 50-110-9263 Supplier Accuflow Systems Supplier No. 9651087
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Platinum-Cured Silicone Tubing, B/T 87, 10' No leachable additives, DOP, or plasticizers; phthalate & latex-free; odorless & nontoxic, fungus, weather, ozone, corona, & radiation-resistant Extreme