Learn More
Accuflow Systems TUBING MFLEX SILICONE #88 10FT
Supplier:  Accuflow Systems 9651088
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, I/P 88, 10 ft. Nontoxic and nonhemolytic with excellent biocompatibility Long pumping life; pump at continuous pressures up to 40 psi Low extractables and low gas
Catalog No. 50-110-8918