Learn More
Accuflow Systems TUBING MFLEX SILICONE #91 10FT
Catalog No. 501108938
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Platinum-Cured Silicone Tubing, B/T 91, 10' No leachable additives, DOP, or plasticizers; phthalate & latex-free; odorless & nontoxic, fungus-resistant. PerfectPosition™ tubing retention ma
Catalog No. 50-110-8938 Supplier Accuflow Systems Supplier No. 9651091
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Platinum-Cured Silicone Tubing, B/T 91, 10' No leachable additives, DOP, or plasticizers; phthalate & latex-free; odorless & nontoxic, fungus-resistant. PerfectPosition™ tubing retention ma