Learn More
Accuflow Systems Masterflex Pump Tubing, Gore STA-PURE PCS, L/S 14, Platinum-cured Silicone
Catalog No. NC1583680
Encompass_Preferred
Masterflex Gore STA-PURE PCS Series L/S 14 pump tubing is a composite peristaltic pump tubing constructed from platinum-cured silicone with a PTFE lattice, engineered for precision fluid transfer in pharmaceutical, bioprocessing, and analytical applications. It provides consistent flow performance, extended pump life, and high chemical resistance for demanding processes.
  • Composite construction of platinum-cured silicone with PTFE lattice for chemical resistance.
  • Low extractables and biocompatible fluid-contact surface for sensitive applications.
  • High burst resistance and durable performance at elevated pressures.
  • Long pump life for reduced maintenance and consistent flow accuracy.
  • Compatible with L/S 14 peristaltic pump heads for precision dosing and transfer.
Catalog No. NC1583680 Supplier Accuflow Systems Supplier No. 9624114
Masterflex Gore STA-PURE PCS Series L/S 14 pump tubing is a composite peristaltic pump tubing constructed from platinum-cured silicone with a PTFE lattice, engineered for precision fluid transfer in pharmaceutical, bioprocessing, and analytical applications. It provides consistent flow performance, extended pump life, and high chemical resistance for demanding processes.
  • Composite construction of platinum-cured silicone with PTFE lattice for chemical resistance.
  • Low extractables and biocompatible fluid-contact surface for sensitive applications.
  • High burst resistance and durable performance at elevated pressures.
  • Long pump life for reduced maintenance and consistent flow accuracy.
  • Compatible with L/S 14 peristaltic pump heads for precision dosing and transfer.